1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperazine 95+%

Supplier: Apollo Scientific
Warning

PC11195-1G PC11195-250MG PC11195-500MG
APOSPC11195-1GEA 348 EUR
APOSPC11195-1G APOSPC11195-250MG APOSPC11195-500MG
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperazine 95+%
1-[4-Fluoro-2-(methylsulphonyl)phenyl]piperazine
Formula: C₁₁H₁₅FN₂O₂S
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD04037948
CAS Number: 849938-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR