1-Fluoropentane 97%

Supplier: Apollo Scientific
Danger

Synonyms: Pentyl fluoride

PC1120-1G PC1120-25G PC1120-5G
APOSPC1120-1GEA 72.5 EUR
APOSPC1120-1G APOSPC1120-25G APOSPC1120-5G
1-Fluoropentane 97%
1-Fluoropentane
Formula: C₅H₁₁F
MW: 90.14 g/mol
Boiling Pt: 62…63 °C
Melting Pt: -80
Density: 0.789 g/cm³ (25 °C)
Flash Pt: –23 °C
Storage Temperature: Ambient
CAS Number: 592-50-7
EINECS: 209-761-5
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR