Potassium-3-(benzyloxy)phenyltrifluoroborate

Supplier: Apollo Scientific

PC11232-1G PC11232-5G
APOSPC11232-1GEA 21.5 EUR
APOSPC11232-1G APOSPC11232-5G
Potassium-3-(benzyloxy)phenyltrifluoroborate
Potassium 3-(benzyloxy)phenyltrifluoroborate
Formula: C₁₃H₁₁BF₃KO
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD04115751
CAS Number: 850623-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR