Pentafluorophenyl pentamethylbenzenesulphonate

Supplier: Apollo Scientific

Synonyms: 2,3,4,5,6-Pentafluorophenyl 2,3,4,5,6-pentamethylbenzenesulphonate

PC11276-1G PC11276-500MG PC11276-5G
APOSPC11276-1GEA 275 EUR
APOSPC11276-1G APOSPC11276-500MG APOSPC11276-5G
Pentafluorophenyl pentamethylbenzenesulphonate
Pentafluorophenyl pentamethylbenzenesulphonate
Formula: C₁₇H₁₅F₅O₃S
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD05975144
CAS Number: 886361-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR