Antimony(V) fluoride
Supplier: Apollo Scientific
|
Warning
|
PC1130-50G
PC1130-10G
APOSPC1130-50GEA
350
EUR
APOSPC1130-50G
APOSPC1130-10G
Antimony(V) fluoride
Antimony (V) fluoride
Formula:
F₅Sb MW: 216,75 g/mol Boiling Pt: 150 °C Melting Pt: 7 °C Density: 2,993 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011219 CAS Number: 7783-70-2 EINECS: 232-021-8 UN: 1732 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...