Chlorofluoroacetic acid 97%

Supplier: Apollo Scientific
Danger

PC1148-5G PC1148-1G
APOSPC1148-5GEA 375 EUR
APOSPC1148-5G APOSPC1148-1G
Chlorofluoroacetic acid 97%
Chlorofluoroacetic acid
Formula: C₂H₂ClFO₂
Boiling Pt: 162 °C
Density: 1,532 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00871521
CAS Number: 471-44-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR