4-Amino-3-fluoropyridine 97%
Supplier: Apollo Scientific
|
Warning
|
PC1163-1G
PC1163-5G
APOSPC1163-1GEA
20.5
EUR
APOSPC1163-1G
APOSPC1163-5G
4-Amino-3-fluoropyridine 97%
4-Amino-3-fluoropyridine
Formula:
C₅H₅FN₂ MW: 112,11 g/mol Melting Pt: 71…79 °C Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234072 CAS Number: 2247-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...