Pyridine-2-sulphonyl Fluoride
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
PyFluor
PC1178-10G
PC1178-1G
PC1178-5G
APOSPC1178-10GEA
304
EUR
APOSPC1178-10G
APOSPC1178-1G
APOSPC1178-5G
Pyridine-2-sulphonyl Fluoride
Pyridine-2-sulphonyl Fluoride
Formula:
C₅H₄FNO₂S MW: 161,15 g/mol Melting Pt: 29…34 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09265513 CAS Number: 878376-35-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...