4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol 97%
Supplier: Apollo Scientific
|
Warning
|
PC1220-5G
PC1220-25G
APOSPC1220-5GEA
248
EUR
APOSPC1220-5G
APOSPC1220-25G
4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol 97%
4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol
Formula:
C₈H₆F₄O MW: 194,13 g/mol Melting Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093260 CAS Number: 79538-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...