Bismuth(V) fluoride 95%
Supplier: Apollo Scientific
PC1230E-10G
PC1230E-25G
PC1230E-5G
APOSPC1230E-10GEA
400
EUR
APOSPC1230E-10G
APOSPC1230E-25G
APOSPC1230E-5G
Bismuth(V) fluoride 95%
Bismuth (V) fluoride
Formula:
BiF₅ MW: 303,97 g/mol Melting Pt: 550 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00015967 CAS Number: 7787-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...