Bis(pentafluorophenyl)dimethylsilane 97%
Supplier: Apollo Scientific
|
Warning
|
PC1234-1G
PC1234-5G
PC1234-25G
APOSPC1234-1GEA
58
EUR
APOSPC1234-1G
APOSPC1234-5G
APOSPC1234-25G
Bis(pentafluorophenyl)dimethylsilane 97%
Bis(pentafluorophenyl)dimethylsilane
Formula:
C₁₄H₆F₁₀Si Boiling Pt: 92…94 °C/1mmHg Melting Pt: 30…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042066 CAS Number: 10536-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...