2-Fluoro-6-(trifluoromethyl)pyridine 97%

Supplier: Apollo Scientific
Warning

PC1264-25G PC1264-5G PC1264-100G
APOSPC1264-25GEA 50.9 EUR
APOSPC1264-25G APOSPC1264-5G APOSPC1264-100G
2-Fluoro-6-(trifluoromethyl)pyridine 97%
2-Fluoro-6-(trifluoromethyl)pyridine
Formula: C₆H₃F₄N
MW: 165,09 g/mol
Boiling Pt: 143 °C
Melting Pt: –15,2 °C
Density: 1,4 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092901
CAS Number: 94239-04-0
EINECS: 428-100-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR