3-Fluoro-1-propanol 97%

Supplier: Apollo Scientific
Danger

PC1281-25G PC1281-100G PC1281-5G
APOSPC1281-25GEA 40.6 EUR
APOSPC1281-25G APOSPC1281-100G APOSPC1281-5G
3-Fluoro-1-propanol 97%
3-Fluoro-1-propanol
Formula: C₃H₇FO
MW: 78,09 g/mol
Boiling Pt: 127,5…128 °C/40mmHg
Density: 1,039 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00039548
CAS Number: 462-43-1
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR