Methyl-2,2-difluoro-3-oxopentanoate

Supplier: Apollo Scientific
Warning

PC1285-25G PC1285-5G
APOSPC1285-25GEA 0 EUR
APOSPC1285-25G APOSPC1285-5G
Methyl-2,2-difluoro-3-oxopentanoate
Methyl-2,2-difluoro-3-oxopentanoate
Formula: C₆H₈F₂O₃
MW: 166,13 g/mol
Boiling Pt: 50 °C/7mmHg
Density: 1,1 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00792443
CAS Number: 196202-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR