3,3,4,4,4-Pentafluorobutan-2-one 97%

Supplier: Apollo Scientific
Warning

PC1288-1G PC1288-5G PC1288-25G
APOSPC1288-1GEA 183 EUR
APOSPC1288-1G APOSPC1288-5G APOSPC1288-25G
3,3,4,4,4-Pentafluorobutan-2-one 97%
3,3,4,4,4-Pentafluorobutan-2-one

  • Solubility in water 7.2%.
  • Surface tension 14.2 dynes/cm.

Formula: C₄H₃F₅O
Boiling Pt: 42 °C
Density: 1,284 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094116
CAS Number: 374-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR