2,3,5,6-Tetrafluorobenzyl alcohol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2,3,5,6-Tetrafluorophenyl)methanol
PC1300-25G
PC1300-1G
PC1300-5G
APOSPC1300-25GEA
29
EUR
APOSPC1300-25G
APOSPC1300-1G
APOSPC1300-5G
2,3,5,6-Tetrafluorobenzyl alcohol 98%
2,3,5,6-Tetrafluorobenzyl alcohol
Formula:
C₇H₄F₄O MW: 180,1 g/mol Boiling Pt: 220…225 °C Melting Pt: 34…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792428 CAS Number: 4084-38-2 EINECS: 443-840-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...