4,4,4-Trifluorobutyronitrile 97%
Supplier: Apollo Scientific
|
Danger
|
PC1301-1G
PC1301-5G
APOSPC1301-1GEA
97.8
EUR
APOSPC1301-1G
APOSPC1301-5G
4,4,4-Trifluorobutyronitrile 97%
4,4,4-Trifluorobutyronitrile
Formula:
C₄H₄F₃N MW: 123.08 g/mol Boiling Pt: 139…140 °C Density: 1.21 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD02183570 CAS Number: 690-95-9 UN: 3273 ADR: 3+6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...