3'-Chloro-4'-fluoroacetanilide 98%
Supplier: Apollo Scientific
Synonyms:
N-(3-Chloro-4-fluorophenyl)acetamide
PC1318-100G
PC1318-25G
PC1318-5G
APOSPC1318-100GEA
125
EUR
APOSPC1318-100G
APOSPC1318-25G
APOSPC1318-5G
3'-Chloro-4'-fluoroacetanilide 98%
3'-Chloro-4'-fluoroacetanilide
Formula:
C₈H₇ClFNO MW: 187,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00018095 CAS Number: 877-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...