1-Nitro-2-(trifluoromethoxy)benzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Trifluoromethoxy)nitrobenzene
PC1348-100G
PC1348-25G
PC1348-5G
APOSPC1348-100GEA
104
EUR
APOSPC1348-100G
APOSPC1348-25G
APOSPC1348-5G
1-Nitro-2-(trifluoromethoxy)benzene
1-Nitro-2-(trifluoromethoxy)benzene
Formula:
C₇H₄F₃NO₃ Boiling Pt: 98 °C/16mmHg Flash Pt: 80 °C Storage Temperature: Ambient |
CAS Number:
1644-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...