2-Chloro-3-fluorotoluene 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Chloro-2-fluoro-6-methylbenzene, 2-Chloro-1-fluoro-3-methylbenzene
PC1360-5G
APOSPC1360-5GEA
127
EUR
APOSPC1360-5G
2-Chloro-3-fluorotoluene 98%
2-Chloro-3-fluorotoluene
Formula:
C₇H₆ClF MW: 144,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08445246 CAS Number: 116850-28-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...