2-Bromo-4-fluoroanisole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-4-fluoro-1-methoxybenzene
PC1397T-100G
PC1397T-250G
PC1397T-25G
APOSPC1397T-100GEA
43.5
EUR
APOSPC1397T-100G
APOSPC1397T-250G
APOSPC1397T-25G
2-Bromo-4-fluoroanisole 97%
2-Bromo-4-fluoroanisole
Formula:
C₇H₆BrFO MW: 205,03 g/mol Boiling Pt: 88…89 °C/7mmHg Density: 1,581 g/cm³ (25 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012014 CAS Number: 452-08-4 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...