3-Bromo-4-fluorobenzaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
PC1399-100G
PC1399-25G
APOSPC1399-100GEA
59.6
EUR
APOSPC1399-100G
APOSPC1399-25G
3-Bromo-4-fluorobenzaldehyde 95%
3-Bromo-4-fluorobenzaldehyde
Formula:
C₇H₄BrFO Boiling Pt: 138…139 °C/2,5mmHg Melting Pt: 31…33 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042186 CAS Number: 77771-02-9 EINECS: 278-764-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...