4-Bromo-2-fluorobenzaldehyde 98%

Supplier: Apollo Scientific
Warning

PC1399F-100G PC1399F-25G
APOSPC1399F-100GEA 58.2 EUR
APOSPC1399F-100G APOSPC1399F-25G
4-Bromo-2-fluorobenzaldehyde 98%
4-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203,01 g/mol
Boiling Pt: 42 °C/19mmHg
Melting Pt: 58…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00143261
CAS Number: 57848-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR