5-Bromo-2-fluorobenzylamine hydrochloride 97%
Supplier: Apollo Scientific
|
Warning
|
PC1423GD-1G
PC1423GD-25G
PC1423GD-5G
APOSPC1423GD-1GEA
67.2
EUR
APOSPC1423GD-1G
APOSPC1423GD-25G
APOSPC1423GD-5G
5-Bromo-2-fluorobenzylamine hydrochloride 97%
5-Bromo-2-fluorobenzylamine hydrochloride
Formula:
C₇H₈BrClFN Melting Pt: 221…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143426 CAS Number: 202865-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...