4-Bromo-2-fluorobiphenyl 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromo-2-fluoro-1,1'-biphenyl
PC1423I-100G
PC1423I-250G
PC1423I-25G
PC1423I-500G
APOSPC1423I-100GEA
40.6
EUR
APOSPC1423I-100G
APOSPC1423I-250G
APOSPC1423I-25G
APOSPC1423I-500G
4-Bromo-2-fluorobiphenyl 97%
4-Bromo-2-fluorobiphenyl
Formula:
C₁₂H₈BrF MW: 251,1 g/mol Boiling Pt: 175 °C/2mmHg Melting Pt: 38…41 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051716 CAS Number: 41604-19-7 EINECS: 255-453-9 UN: 3152 ADR: 9,II |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...