4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline 95%

Supplier: Apollo Scientific
Warning

PC1443-1G PC1443-250MG
APOSPC1443-1GEA 30.6 EUR
APOSPC1443-1G APOSPC1443-250MG
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline 95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline
Formula: C₁₂H₈ClF₃N₂O
MW: 288,66 g/mol
Melting Pt: 85…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00170933
CAS Number: 71422-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR