3-Amino-5-(trifluoromethyl)-1H-indazole

Supplier: Apollo Scientific
Danger

Synonyms: 5-(Trifluoromethyl)-1H-indazol-3-amine

PC1447-1G PC1447-5G
APOSPC1447-1GEA 46.4 EUR
APOSPC1447-1G APOSPC1447-5G
3-Amino-5-(trifluoromethyl)-1H-indazole
3-Amino-5-(trifluoromethyl)-1H-indazole
Formula: C₈H₆F₃N₃
MW: 201,15 g/mol
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD01719740
CAS Number: 2250-53-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR