5-Bromo-2-fluorotoluene 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromo-1-fluoro-2-methylbenzene
PC1460-100G
PC1460-500G
APOSPC1460-100GEA
40.6
EUR
APOSPC1460-100G
APOSPC1460-500G
5-Bromo-2-fluorotoluene 97%
5-Bromo-2-fluorotoluene
Formula:
C₇H₆BrF MW: 189,03 g/mol Boiling Pt: 95 °C/50mmHg Density: 1,486 g/cm³ (25 °C) Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000343 CAS Number: 51437-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...