1-Chloro-2,3-difluoro-6-nitrobenzene 96%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-3,4-difluoronitrobenzene

PC1497-1G PC1497-5G PC1497-25G
APOSPC1497-1GEA 21.9 EUR
APOSPC1497-1G APOSPC1497-5G APOSPC1497-25G
1-Chloro-2,3-difluoro-6-nitrobenzene 96%
1-Chloro-2,3-difluoro-6-nitrobenzene
Formula: C₆H₂ClF₂NO₂
MW: 193,5354 g/mol
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD06658262
CAS Number: 169468-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR