Gatifloxacin

Supplier: Apollo Scientific

Synonyms: 1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

PC1528-100G PC1528-10G PC1528-25G
APOSPC1528-100GEA 575 EUR
APOSPC1528-100G APOSPC1528-10G APOSPC1528-25G
Gatifloxacin
Gatifloxacin
Formula: C₁₉H₂₂FN₃O₄
MW: 375,40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00895399
CAS Number: 112811-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR