5-Bromo-4,4,5,5-tetrafluoropentanoic acid 95%
Supplier: Apollo Scientific
|
Danger
|
PC1567J-5G
PC1567J-25G
APOSPC1567J-5GEA
212
EUR
APOSPC1567J-5G
APOSPC1567J-25G
5-Bromo-4,4,5,5-tetrafluoropentanoic acid 95%
5-Bromo-4,4,5,5-tetrafluoropentanoic acid
Formula:
C₅H₅BrF₄O₂ Melting Pt: 33…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153717 CAS Number: 234443-22-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...