4-Bromo-2-(trifluoromethoxy)aniline 99%

Supplier: Apollo Scientific
Danger

PC1593K-100G PC1593K-25G PC1593K-500G
APOSPC1593K-100GEA 69.6 EUR
APOSPC1593K-100G APOSPC1593K-25G APOSPC1593K-500G
4-Bromo-2-(trifluoromethoxy)aniline 99%
4-Bromo-2-(trifluoromethoxy)aniline
Formula: C₇H₅BrF₃NO
Boiling Pt: 98…100 °C/15mmHg
Melting Pt: 29…30 °C
Density: 1,691 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00179338
CAS Number: 175278-09-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR