2-Bromo-3-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
|
Danger
|
PC1598E-25G
PC1598E-5G
APOSPC1598E-25GEA
120
EUR
APOSPC1598E-25G
APOSPC1598E-5G
2-Bromo-3-(trifluoromethyl)pyridine 97%
2-Bromo-3-(trifluoromethyl)pyridine
Formula:
C₆H₃BrF₃N MW: 226 g/mol Boiling Pt: 76 °C/8mmHg Melting Pt: 37…41 °C Flash Pt: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153084 CAS Number: 175205-82-0 UN: 2926 ADR: 4.1+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...