3-Fluoro-5-isobutoxybenzeneboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Fluoro-5-(isobutoxy)benzeneboronicacid
PC1608-5G
PC1608-25G
APOSPC1608-5GEA
139
EUR
APOSPC1608-5G
APOSPC1608-25G
3-Fluoro-5-isobutoxybenzeneboronic acid 98%
3-Fluoro-5-isobutoxybenzeneboronic acid
Formula:
C₁₀H₁₄BFO₃ MW: 212,03 g/mol Melting Pt: 130…134 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363783 CAS Number: 850589-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...