2-Chloro-6-fluorophenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2-Chloro-6-fluorophenyl)boronic acid, (2-Chloro-6-fluorophenyl)metaboric acid, 2-Chloro-6-fluorobenzeneboronic acid, Dihydroxy(2-chloro-6-fluorophenyl)borane
PC1633-100G
PC1633-25G
APOSPC1633-100GEA
401
EUR
APOSPC1633-100G
APOSPC1633-25G
2-Chloro-6-fluorophenylboronic acid 98%
2-Chloro-6-fluorophenylboronic acid
Formula:
C₆H₅BClFO₂ MW: 174,37 g/mol Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039892 CAS Number: 313545-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...