4-Chloro-2-fluoro-5-nitrobenzoic acid 98%

Supplier: Apollo Scientific
Warning

PC1692-1G PC1692-25G PC1692-5G PC1692-100G
APOSPC1692-1GEA 14.7 EUR
APOSPC1692-1G APOSPC1692-25G APOSPC1692-5G APOSPC1692-100G
4-Chloro-2-fluoro-5-nitrobenzoic acid 98%
4-Chloro-2-fluoro-5-nitrobenzoic acid
Formula: C₇H₃ClFNO
MW: 219,55 g/mol
Melting Pt: 146…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115725
CAS Number: 35112-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR