6-Fluorooxindole 97%

Supplier: Apollo Scientific
Warning

PC1736-5G PC1736-1G
APOSPC1736-5GEA 98.9 EUR
APOSPC1736-5G APOSPC1736-1G
6-Fluorooxindole 97%
6-Fluorooxindole
Formula: C₈H₆FNO
Melting Pt: 132…136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047212
CAS Number: 56341-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR