Chlorodifluoroacetic acid 98%

Supplier: Apollo Scientific
Danger

Synonyms: Difluorochloroacetic acid

PC1740-100G PC1740-25G
APOSPC1740-100GEA 66.7 EUR
APOSPC1740-100G APOSPC1740-25G
Chlorodifluoroacetic acid 98%
Chlorodifluoroacetic acid

On treatment with base difluorocarbene can be generated: Tetrahedron Lett., 21, 1047 (1980), Yoshida et al., J. Fluorine Chem., 1994, 68, 33-38.

Formula: C₂HClF₂O₂
MW: 130,48 g/mol
Boiling Pt: 121…122 °C
Melting Pt: 20…24 °C
Density: 1,54 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004176
CAS Number: 76-04-0
EINECS: 200-928-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR