5-(Trifluoromethyl)oxindole 98%
Supplier: Apollo Scientific
|
Warning
|
PC1742-100MG
PC1742-1G
PC1742-5G
APOSPC1742-100MGEA
102
EUR
APOSPC1742-100MG
APOSPC1742-1G
APOSPC1742-5G
5-(Trifluoromethyl)oxindole 98%
5-(Trifluoromethyl)oxindole
Formula:
C₉H₆F₃NO Melting Pt: 182…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179602 CAS Number: 71293-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...