3-Carboxy-5-fluorophenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Carboxy-5-fluorobenzeneboronic acid, 3-Borono-5-fluorobenzoic acid

PC1770-5G
APOSPC1770-5GEA 127 EUR
APOSPC1770-5G
3-Carboxy-5-fluorophenylboronic acid 98%
3-Carboxy-5-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183,93 g/mol
Melting Pt: 112…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363791
CAS Number: 871329-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR