3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 96%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-5-methoxycarbonylphenylboronic Acid, 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

PC1774-5G
APOSPC1774-5GEA 149 EUR
APOSPC1774-5G
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 96%
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197,96 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363749
CAS Number: 871329-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR