4-Fluoro-3-(trifluoromethyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid

PC1779-5G PC1779-1G PC1779-25G
APOSPC1779-5GEA 24.8 EUR
APOSPC1779-5G APOSPC1779-1G APOSPC1779-25G
4-Fluoro-3-(trifluoromethyl)phenylboronic acid 98%
4-Fluoro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MW: 207,92 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD07363787
CAS Number: 182344-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR