1-(2-Chloroethyl)-4-fluorobenzene 98%

Supplier: Apollo Scientific
Warning

PC1812J-1G PC1812J-5G PC1812J-25G
APOSPC1812J-1GEA 29 EUR
APOSPC1812J-1G APOSPC1812J-5G APOSPC1812J-25G
1-(2-Chloroethyl)-4-fluorobenzene 98%
1-(2-Chloroethyl)-4-fluorobenzene
Formula: C₈H₈ClF
MW: 158,6 g/mol
Boiling Pt: 98…99 °C/26mmHg
Density: 1,507 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00155000
CAS Number: 332-43-4
EINECS: 206-364-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR