3-Chloro-4-fluorobenzaldehyde 96%
Supplier: Apollo Scientific
|
Warning
|
PC1828G-100G
PC1828G-25G
APOSPC1828G-100GEA
80
EUR
APOSPC1828G-100G
APOSPC1828G-25G
3-Chloro-4-fluorobenzaldehyde 96%
3-Chloro-4-fluorobenzaldehyde
Formula:
C₇H₄ClFO MW: 158,56 g/mol Boiling Pt: 66…72 °C/2mmHg Melting Pt: 25…30 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011735 CAS Number: 34328-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...