α-Bromo-3-chloro-2-fluorotoluene 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-3-chloro-2-fluorobenzene
PC1864G-5G
PC1864G-100G
PC1864G-25G
APOSPC1864G-5GEA
21.9
EUR
APOSPC1864G-5G
APOSPC1864G-100G
APOSPC1864G-25G
α-Bromo-3-chloro-2-fluorotoluene 97%
α-Bromo-3-chloro-2-fluorotoluene
Formula:
C₇H₅BrClF MW: 223,47 g/mol Boiling Pt: 125…126 °C/15mmHg Melting Pt: 36…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042503 CAS Number: 85070-47-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...