1-Chloro-3-fluoro-2-propanol 96%
Supplier: Apollo Scientific
|
Warning
|
PC1868M-1G
PC1868M-25G
PC1868M-5G
APOSPC1868M-1GEA
114
EUR
APOSPC1868M-1G
APOSPC1868M-25G
APOSPC1868M-5G
1-Chloro-3-fluoro-2-propanol 96%
1-Chloro-3-fluoro-2-propanol
Formula:
C₃H₆ClFO MW: 112,53 g/mol Boiling Pt: 153…156 °C Density: 1,3 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00045284 CAS Number: 453-11-2 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...