3-Chloro-4-fluorophenol 97%
Supplier: Apollo Scientific
|
Danger
|
PC1880D-25G
PC1880D-1G
PC1880D-5G
APOSPC1880D-25GEA
94.5
EUR
APOSPC1880D-25G
APOSPC1880D-1G
APOSPC1880D-5G
3-Chloro-4-fluorophenol 97%
3-Chloro-4-fluorophenol
Formula:
C₆H₄ClFO MW: 146,55 g/mol Boiling Pt: 103…104 °C/11mmHg Melting Pt: 38…43 °C Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002257 CAS Number: 2613-23-2 EINECS: 220-042-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...