2-Chloro-6-fluorophenylacetonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
PC1884-10G
PC1884-5G
APOSPC1884-10GEA
238
EUR
APOSPC1884-10G
APOSPC1884-5G
2-Chloro-6-fluorophenylacetonitrile 98%
2-Chloro-6-fluorophenylacetonitrile
Formula:
C₈H₅ClFN MW: 169,59 g/mol Boiling Pt: 85 °C/2mmHg Melting Pt: 41…42 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001900 CAS Number: 75279-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...