2-Chloro-5-fluorotoluene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Chloro-4-fluoro-2-methylbenzene
PC1900-100G
PC1900-25G
PC1900-5G
APOSPC1900-100GEA
69.6
EUR
APOSPC1900-100G
APOSPC1900-25G
APOSPC1900-5G
2-Chloro-5-fluorotoluene
2-Chloro-5-fluorotoluene
Formula:
C₇H₆ClF Boiling Pt: 156…157 °C Density: 1,2 g/cm³ (25 °C) Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040853 CAS Number: 33406-96-1 EINECS: 251-508-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...