5-Chloro-2-fluorotoluene 99%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chloro-1-fluoro-2-methylbenzene

PC1930-25G PC1930-5G PC1930-100G PC1930-500G
APOSPC1930-25GEA 23.2 EUR
APOSPC1930-25G APOSPC1930-5G APOSPC1930-100G APOSPC1930-500G
5-Chloro-2-fluorotoluene 99%
5-Chloro-2-fluorotoluene
Formula: C₇H₆ClF
MW: 144,58 g/mol
Boiling Pt: 156 °C
Density: 1,188 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00018504
CAS Number: 452-66-4
EINECS: 207-204-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR